Sodium decyl sulphate, Supelco®

Leverandør: Merck

Synonymer: Decyl sodium sulphate, Decyl sodium sulfate, Decyl sulphuric acid salt, Decyl sulfuric acid salt

M-Clarity™ quality level = MQ100

71443-10G
71443-10GEA 5630 NOK
71443-10G
Sodium decyl sulphate, Supelco®
Sodium decyl sulphate
Formel: C₁₀H₂₁NaO₄S
Tetthet: 1,207 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00041881
CAS-nummer: 142-87-0
EINECS: 205-568-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d82a954ab65491ffc3b7350e51258fadc07152a2&vid=ed551da13fbd661e7ea1751df0785a5ba4b17c4c&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR