Staining racks
Leverandør: DIXON SCIENTIFIC
406019000
720-0096EA 4475 NOK
720-0096
Staining racks
Stativer for rør Fargestativer
Comprises two brass end-plates fitted with levelling screws and clips.
  • For holding two glass rods with diameter of ∼8 to 9 mm
Bestillingsinformasjon: Rods are not included.