Swabs, Steriswabs®
Leverandør: MEDICAL WIRE EQUIPMENT
MW720
720-0142EA 2650 NOK
720-0142
Swabs, Steriswabs®
Svabere Svabere for prøvetaking
Swabs with 45 mm breakpoint, dry or pre-moistened with sterile diluent.
  • Available in labelled tube or peel pouches
  • Triple wrapped and radiation sterilised
Designed for effective monitoring of sterility and contamination in clinically clean or sterile work areas.
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904