Brilliant green, crystals

Leverandør: RAL
Warning

Synonymer: Ethyl Green, CI 42040, Basic Green 1, N-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulphate, Diamond Green, Malachite Green G, Emerald Green, Astradiamant green GX, Solid Green JO, Brilliant Green hydrogen sulphate

317800-0100
720-0185EA 2225 NOK
720-0185
Brilliant green, crystals
Brilliant green

pH indicator

Formel: C₂₇H₃₄N₂O₄S
MW: 482.65 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011880
CAS-nummer: 633-03-4
EINECS: 211-190-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR