Lys Grønn SF Gulaktig

Leverandør: RAL

Synonymer: Dihydrogen (ethyl)[4-[p-[ethyl(m-sulphonatobenzyl)amino]-p'-sulphonatobenzhydrylene]cyclohexa-2,5-dien-1-ylidene](m-sulphonatobenzyl)ammonium disodium salt, Acid Green 5, Light Green 2G, S or 2GN, CI 42095, Sodium 3-((-ethyl(-4-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-sulphonatophenyl)methylene)cyclohexa-2,5-dien-1-ylidene)ammonio)methyl)benzenesulphonate, Food Green n°2, Acid Green F, Fast acid Green N, CN DcL-30

320770-0025
720-0335EA 4175 NOK
720-0335
Lys Grønn SF Gulaktig
Lys Grønn SF Gulaktig

Light green SF is a bluish green. it is used to stain bacteria , algae and yeast in various methods

Formel: C₃₇H₃₄N₂Na₂O₉S₃
MW: 792.86 g/mol
Smeltepunkt: 288…289 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012121
CAS-nummer: 5141-20-8
EINECS: 225-906-5
Merck Indeks: 13,05506
IVDLogo

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR