Gram-Hücker R Kit To differentiate Gram-positive from Gram-negative Bacteria

Leverandør: RAL

361520-0000
720-0350EA 915 NOK
720-0350
Gram-Hücker R Kit To differentiate Gram-positive from Gram-negative Bacteria
Gram-Hücker R Kit

Gram-Hücker staining is based on the same principle than the Gram-Nicolle staining. The structure of the bacterial walls and their permeability to alcohol help differentiate the Gram-negative bacteria from the Gram-positive ones. In this technique, Carbolic Gentian Violet is replaced by Crystal Violet Oxalate (that does not require filtration) and Ziehl-Carbolic Fuchsin by Safranin.


Components - Name (ref) Packing
CRISTAL VIOLET OXALATE (Ref 361490) 240 ml (Other packagings 1000, 2500 ml)
LUGOL PVP STABILIZED SOLUTION (Ref 367400) 240 ml (Other packagings 1000, 2500 ml)
DIFFÉRENCIATEUR FAST (Ref 361510) 240 ml (Other packagings 1000, 2500 ml)
SAFRANIN (Ref 361500) 240 ml (Other packagings 1000 ml, 2500 ml)

IVDLogo

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR