HistoPerls Kit Highlighting hemosiderin, pathological pigment (ferric salts)
Leverandør: RAL
361850-0000
720-0359EA 2675 NOK
720-0359
HistoPerls Kit Highlighting hemosiderin, pathological pigment (ferric salts)
HistoPerls Kit
In an acid environment, ferric ions react with Potassium Ferrocyanide to form a precipitate: Ferric Ferrocyanide (or Prussian Blue), that shows the presence of the pathological pigment, Haemosiderin.This pigment is found in the liver and in bone marrow in diseases such as Haemochromatosis, Cirrhosis and some anemia.
Components - Name (ref) Packing
NUCLEAR RED IN SOLUTION (ref 320910) 100 ml
ACID BUFFER AND POTASSIUM FERROCYANIDE (Ref 361955) 5 ml (Other packagings 1000 ml)
IVDLogo