Scintillation vials, Snaptwist®
Leverandør: Simport Scientific
S207 S207-5
720-0494EA 1800 NOK
720-0494 720-0495
Scintillation vials, Snaptwist®
Vialer
General purpose vials made from PP and HDPE for liquid scintillation and gamma counting, chromatography, sample storage and culturing. Can be used with the LKB and Packard Varisette counters.
  • Easy removal of caps (¼ turn-twist) helps to eliminate the danger of spillage
  • A built-in positive lock prevents the cap from popping off
  • Shoulderless vials feature a full width opening of 12,5 mm