Erythrosin B disodium salt

Leverandør: RAL
Warning

Synonymer: C.I. 45430

350150-0100
720-1142EA 3275 NOK
720-1142
Erythrosin B disodium salt
Erythrosin B disodium salt

Indicator Ph and laboratory reagent use in histo-cytology

Formel: C₂₀H₆I₄Na₂O₅
MW: 879.86 g/mol
MDL nummer: MFCD00144257
CAS-nummer: 16423-68-0
EINECS: 240-474-8
IVDLogo

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR