Phloxin B for histology, for cytology

Leverandør: RAL

Synonymer: CI 45410

361470-0025
720-1159EA 1550 NOK
720-1159
Phloxin B for histology, for cytology
Phloxin B
Formel: C₂₀H₂Br₄Cl₄Na₂O₅
MW: 829.6344 g/mol
MDL nummer: MFCD00070627
CAS-nummer: 18472-87-2
EINECS: 242-355-6
IVDLogo

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR