Quick TB Kit Cold Staining
Leverandør: RAL
361560-0000
720-1166EA 1000 NOK
720-1166
Quick TB Kit Cold Staining
Quick TB Kit
Staining with Quick TB Kit allows to detect mycobacteria, also called A.F.B (Acid Alcohol Fast Bacilli) through a cold and fast staining technique. Carbolic Fuchsin RAL stains mycobacteria pink. Armand Solution stains the background of the preparation blue.
Components - Name (ref) Packing
CARBOLIC FUCHSIN RAL (Ref 365240) 125 ml (Other packaging 1000 ml)
ARMAND STAIN (Ref 360100) 125 ml (Other packaging 1000 ml)
IVDLogo