Hemaperls kit for haematology
Leverandør: RAL
362800-0000
720-1184EA 2675 NOK
720-1184
Hemaperls kit for haematology
Hemaperls kit for haematology
Perls Staining for the Detection of Haemosiderin (Ferric salts - Prussian Blue)
In an acid environment, ferric ions react with potassium ferrocyanide what leads to a precipitate called Ferric Ferrocyanide (Prussian blue). It points out the presence of a pathological pigment: Haemosiderin.
IVDLogo