Vita Eosin Kit, eosin-nigrosin Spermogram : Differentiation between dead and living spermatozoa
Leverandør: RAL
380420-0000
720-1205EA 1375 NOK
720-1205
Vita Eosin Kit, eosin-nigrosin Spermogram : Differentiation between dead and living spermatozoa
Vita Eosin Kit, eosin-nigrosin
The spermogram helps assess the percentage of dead and living spermatozoa. Only dead spermatozoa stain pink by Eosin. Living spermatozoa remain colourless, except their outlines stain dark blue by Nigrosin.
Components - Name (ref) Packing
EOSIN IN SOLUTION (Ref 363200) 100 ml (Other packagings 1000 ml, 2500 ml)
NIGROSIN IN SOLUTION (ref 363210) 100 ml (Other packagings 1000 ml, 2500 ml)
IVDLogo