Multicassettes for drug testing in urine
Leverandør: SURESCREEN
6MC1
720-1239EA 2875 NOK
720-1239
Multicassettes for drug testing in urine
Narkotester
The multicassette tests for up to 12 several drugs simultaneously, and is very easy to use. The urine sample is added to the sample well with the pipette that comes with the kit, and resuts are read at 3 minutes. Tests are individually wrapped in a foil pouch, to ensure the high-quality of the product is retained throughout the shelf-life.
  • Test for up to 12 drug types simultaneously
  • Easy to perform test routine
  • Clear results within 3 minutes
  • Greater than 99% accuracy
  • Wide range of test combinations available
  • Pipette provided for easy sampling
  • Detects