Urine collection cup with screw lid
Leverandør: SURESCREEN
STDCUP/TEMP
720-1333EA 17 NOK
720-1333
Urine collection cup with screw lid
Diagnostiske systemer Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
Urine collection cup with screw lid and temperature strip to show validity of sample. Suitable for use with all types of urine drugs of abuse test, apart from the integrated cup.
  • Designed to accommodate all versions of multipanel test