Cytology funnels, MEGA CytoSep™, for SHANDON Cytospin® 4 cytocentrifuge
Leverandør: Simport Scientific
M965-40
720-1867EA 23575 NOK
720-1867
Cytology funnels, MEGA CytoSep™, for SHANDON Cytospin® 4 cytocentrifuge
Trakter Cytologitrakter
The CytoSep™ MEGA funnel provides an easy, efficient and cost effective method of producing high quality thin layer slide preparations. It eliminates the need for stainless steel slide clips, offering laboratory workflow improvements. It is dedicated for convenient preparation of larger volume samples. This larger funnel is quick and easy to remove. The large rectangular cell deposition area of 22×14,75 mm (325 mm²) provides for up to 12 times the sample volume (6 ml) of the single CytoSep™ cytology funnel. It produces more cost-effective thin layer preparations when compared to other thinMore Product Information