VWR®, manipulasjonssikker beholder for transport av biologiske prøver
Leverandør: VWR Collection
720-1987EA 6350 NOK
720-1987
VWR®, manipulasjonssikker beholder for transport av biologiske prøver
Prøvecontainere Beholdere, generelt bruk
Container for transport of biological samples.
  • Disposable, with tamper-proof seal: Once closed the tube seal must be broken to access the contents
  • Leakproof to prevent accidental outflow of biological fluids that guarantees the safety of transportation personnel
  • Transparent and shock-proof for visual inspection of contents and for ensuring their integrity
Complete with test tube holder made of absorbent material holding 4×16 mm tubes, 4×12 mm tubes and 2×30 mm tubes at the same time, the test tubes are, therefore, protected if accidentally dropped,More Product Information