Blood gas capillary tubes, VITREX®
Leverandør: Vitrex Medical A/S
182313 182413 182513 185013 183313 182013 182613
720-2296EA 980 NOK
720-2296 720-2297 720-2298 720-2299 MODU183313 MODU182013 MODU182613
Blood gas capillary tubes, VITREX®
Pipetter Mikrokapillærer
These disposable capillary tubes for blood gas analyses are made from heavy walled glass.
  • Available non-heparinised, with sodium or lithium heparinisation
  • Careful fire polishing, heavy walled tubes and robust end-caps secure proper function and reduce operating risk
  • Both ends melted
Shelf life: 4 years.