Slide wallets, FiloSlide™
Leverandør: CELLPATH
WEP-9902-00A
720-2347EA 14700 NOK
720-2347
Slide wallets, FiloSlide™
Objektglassholdere
These slide folders feature 21 compartments for 76×26, 50×76 and 76×76 mm slides.
  • Hanging system
  • Storage in conventional filing cabinets