VWR® W10, Slide Warmer/Dryer
Leverandør: VWR Collection
720-2684EA 0 NOK
720-2684 720-2422
VWR® W10, Slide Warmer/Dryer
Varmeplate for objektglass
The W10 heating plate, with its sleek compact design, accommodates two rows of up to 40 slides in total. The heating plate is very useful as a stretch bench for mounting, stretching and drying paraffin sections on slides, fixing bacterial smears, and heating fluid samples in Petri dishes. The black anodised surface provides an excellent visual contrast. The working plate is made of solid 8 mm aluminium with black anodised surface and ensures a constant, homogenous temperature over the entire heating surface. The touch display and switch are protected from water and paraffin. The unit hasMore Product Information