VWR® W20, Histology water bath
Leverandør: VWR Collection
720-2685EA 0 NOK
720-2685 720-2423
VWR® W20, Histology water bath
Bad Vevsbad
The reliable W20 histology water bath is ideal for removing wrinkles and distortions in paraffin wax sections which are created during sectioning. The unit's low profile is ergonomic, provides a clear view, and allows safe handling of sections. The indirectly-heated flat areas at the front and back of the bath provide a suitable space for drying up to 20 slides. Touch-temperature adjustment and power On/Off switch, with a control light for 'On' mode, are conveniently located at the front of the unit and protected from water.
  • Electronic touch display
  • Lightweight, compact and efficient