Single use needles for blood sampling, Sterican®
Leverandør: B. Braun
4665465
720-2533EA 340 NOK
720-2533
Single use needles for blood sampling, Sterican®
Sprøytenåler
Thin walled needles made of stainless chromium-nickel steel with extremely smooth surface and light silicone coating.
  • Special long/short bevel facets for minimal puncture pain
  • Transparent Luer-Lock fitting made from PP
  • Colour code conform to ISO 6009, latex and PVC free
Specially designed for blood sampling applications.
Sertifiseringer: ISO 7864 and DIN 13097.
Forpakning: In a dispenser box.