Cell washer, CW 3
Leverandør: Thermo Scientific
75007405 75007406
720-2858EA 0 NOK
720-2858 521-2652
Cell washer, CW 3
Sentrifuger Sentrifuger, bordmodell
This compact cell washer is designed to provide thorough, reproducible blood cell washing, up to 24 tubes per run.
  • Fully automated cell washing cycle with pre-set programs saves time
  • Manual mode available to wash the cells step-by-step, allowing closed monitoring of washing process
  • Condition-setting panel allows more flexible protocols to optimise washing performance
  • User-friendly operation with keypad control and easy-to-read digital displays
  • Automatic alerts indicate if the saline level is low, and when a run is complete
The CW 3 supports the mostMore Product Information