VWR®, Prepared Slides: General Biology
Leverandør: VWR Collection
720-2924EA 1075 NOK
720-2924
VWR®, Prepared Slides: General Biology
Objektglass Prepared Slides
Set of prepared slides for educational purposes, to better understand general biology.
  • Unicellular animal i.e. amoeba or other
  • Paramecium (w.b.)
  • Hydra (w.b.)
  • Daphnia (w.b.)
  • Lumbricus, earthworm (c.s.)
  • House fly mouth (w.b.)
  • Honey bee mouth (w.b.)
  • Honey bee hind leg
  • Three bacteria smears
  • Simple flat epitelium (w.b.)
  • Skeletal muscle (l.s. and c.s.) human blood smear
  • Frog blood smear
  • Lung section
  • Artery and vein (c.s.)
  • Human skin section through hair follicle
  • Spirogyra conjugation (w.b.)
  • Volvox (w.b.)
  • Moss (w.b.)
  • Mitosis, onion root tip cell (s.)
  • Monocot and