Chromium standard for ICP-MS, TraceCERT®, Supelco®
Leverandør: Merck
72995-100ML
72995-100MLEA 3175 NOK
72995-100ML
Chromium standard for ICP-MS, TraceCERT®, Supelco®
Standarder Kationstandarder Chromium Standards
Prepared with (NH₄)²Cr₂O₇.
Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034