Dual-gel electrophoresis system, Owl™ P10DS

Leverandør: THERMO OWL SCIENTIFIC
P10DS-1
730-0263EA 45350 NOK
730-0263
Dual-gel electrophoresis system, Owl™ P10DS
Elektroforesesystemer Vertikal gelelektroforesesystemer
The Owl™ P10DS dual gel system casts and runs one or two 20×20 cm gels in the upper buffer chamber. The larger size cassette allows the maximum number of samples and separation length. The system is ideal for second dimension protocols of 2-D electrophoresis applications.

  • Optimal for large sample numbers that need extended separation and mobility shift assays

Gel W×H: 200×200 mm
Footprint W×H×D: 310×230×165 mm
Running buffer volume: 1150 ml (upper chamber 650 ml, lower chamber 500 ml)

Leveringsinformasjon: Complete system includes upper bufferMore Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR