VWR® Imager2Imager CHEMI Premium, Gel Documentation System
Leverandør: VWR Collection
730-1458EA 0 NOK
730-1458 730-1469
VWR® Imager2Imager CHEMI Premium, Gel Documentation System
Fotodokumentasjonssystemer
VWR® Imager gel documentation and analysis systems are available in configurations suitable for fluorescence and chemiluminescence applications. Both models are fitted with a digital CCD camera (3,8 or 4 MP) which utilises the latest USB technology. The standard system features a 12-bit CCD camera that can be software modified to 16-bit. The CHEMI Premium system is equipped with a true 16-bit advanced cooled CCD camera able to capture images of a wide range of chemiluminescent samples using common substrates.
  • Modular system can be tailored to meet specific user requirements
  • Automated
More Product Information
Bestill nå
SPESIFIKASJONER
Modell VWR® Imager 2 VWR® Imager CHEMI Premium
Environmentally Preferable
Maks. gelstørrelse, med hette 255×210 mm 305×227 mm
B×D×H 570×450×840 mm 570×450×840 mm