VWR® Basic, Gel Documentation System
Leverandør: VWR Collection
730-1470EA 44475 NOK
730-1470
VWR® Basic, Gel Documentation System
Fotodokumentasjonssystemer
The VWR® Basic gel documentation system offers reproducible, high resolution digital images for gel documentation. This entry level system is easy to use and allows real time images to be captured and viewed directly on a PC or laptop (not supplied). The VWR® Basic system can be illuminated using transillumination from UV, white or blue light. These flexible lighting options make the VWR® Basic suitable for capturing and viewing images from a wide range of samples.
  • High resolution digital camera with 18 MP resolution produces high quality images with