VWR® CHEMI only, Chemiluminescence Imaging System
Leverandør: VWR Collection
730-1471EA 0 NOK
730-1471
VWR® CHEMI only, Chemiluminescence Imaging System
Fotodokumentasjonssystemer
The VWR® CHEMI only system is dedicated to chemiluminescence imaging. The system is built for high performance automation and features a next generation, high quantum efficiency CCD camera for even greater sensitivity. A very simple set-up process means a single click will automatically capture a quality image of any Western blot.
  • Configured for maximum sensitivity to ensure even the faintest band on a blot can be captured
  • High quantum efficiency camera (73% @ 425 nm) is very sensitive to low level light emissions
  • Small footprint - takes up minimal bench