Data storage software, GeneDirectory
Leverandør: Syngene (a division of the Synoptics group)
GENEDTORY-DB
730-2832EA 23600 NOK
730-2832
Data storage software, GeneDirectory
Programvare Laboratoriesoftware
GeneDirectory is a fast data storage and retrieval system designed for large-scale genomic fingerprinting studies, cluster analysis, VNTR analysis, genotyping and RFLP. Used in conjunction with the GeneTools gel analysis package, GeneDirectory can save hours of repetitive work by automatically comparing thousands of bands on different gels.
  • Complete user control with the potential for creating an unlimited number of databases, all with ‘free form’ user input for folder, experiments, gels and tracks
  • Able to handle large quantities of data with methods for