Electrofusion buffer, BTXpress Cytofusion® Medium C
Leverandør: BTX
47-0001
731-0348EA 6700 NOK
731-0348
Electrofusion buffer, BTXpress Cytofusion® Medium C
Nukleinsyre reagenser Transformasjonsreasenser
BTXpress Cytofusion® Medium C is an advanced electrofusion buffer designed especially for hybridoma production. The low conductivity buffer is specially formulated to minimise cell turbulence during cell alignment and heating during electrofusion, for robust cell fusion efficiency and high cell viability. BTXpress Cytofusion® Medium C is sterile filtered from the highest quality non animal, medical grade reagents.