Freezer storage boxes for 0,2 ml PCR tubes, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
F188370000
731-0703EA 2200 NOK
731-0703
Freezer storage boxes for 0,2 ml PCR tubes, SP Bel-Art
Bokser Kryobokser
These storage boxes are designed to fit in a standard 2" freezer racks. This 144-place PCR tube freezer box will hold individual and/or strip 0,2 ml PCR tubes.
  • Designed for ULT freezer storage at temperatures down to −86 °C
  • Durable and impervious to moisture
  • Clear polycarbonate lid and base and alpha-numeric index make it easy to identify samples within the box
  • Two boxes may be stacked to fit together in one space in a 2" freezer storage rack if using flat-capped PCR tubes