PCR box/rack
Leverandør: Brand
781362
731-1251EA 1050 NOK
731-1251
PCR box/rack
Stativer for rør Tube Racks
PP, assorted colours (red, yellow, green, purple, blue).
  • Can be stacked without lids
  • Able to withstand temperatures from −80 to +121 °C
Suitable for sample preparation, for keeping and storing 0,2 ml single tubes, 8-strips and 12-strips, and 96-well PCR plates.