Electroporation Cuvettes Plus
Leverandør: BTX
45-0124 45-0125 45-0126 45-0134
732-0020EA 4225 NOK
732-0020 732-0021 732-0023 BTXI45-0134
Electroporation Cuvettes Plus
Kyvetter Elektroporasjonskyvetter
High quality disposable cuvettes for transfection available in three sizes and compatible with electroporation systems.
  • Transfer pipette for manipulation of samples and gamma irradiated together for sterility
  • Polished aluminium electrodes in the cuvettes lowers arcing frequency
  • Cuvettes (without cap) are autoclavable
Kyvettelokkene er runde for enkel fjerning med én hånd og er fargekodet for rask identifikasjon.