VWR® 96-brønns dypbrønnsplater med automatiseringsspor
Leverandør: VWR Collection
732-0585EA 1025 NOK
732-0585
VWR® 96-brønns dypbrønnsplater med automatiseringsspor
Mikroplater Deepwell mikroplater
Heavy polypropylene deep well plates feature square wells, molded alphanumeric labeling, and smooth interior surfaces to ensure that all the samples can be pipetted.
  • Alphanumeric indexing
  • Square well with V-bottom design
  • Autoclavable
  • Freezable to –80 °C (–112 °F)
VWR® deep-well plates feature printed alphanumeric labeling and smooth interior surfaces to ensure easy pipetting. Plates and mats are free of RNase, DNase and human DNA (lot-by-lot certificates available).
Applications include HTS, combinatorial chemistry, library applications (compound