VWR®, Blotting Pads, Grade 703

Leverandør: VWR Collection
732-0591EA 4500 NOK
732-0591 732-0593 732-0605 732-0596 732-0603
VWR®, Blotting Pads, Grade 703
Blottingpapir
Ideal for use as wicks in protein and nucleic acid blotting. Provides a uniform flow of buffer through the gel to the transfer membrane in a blotting sandwich. Also suitable for use in removing gels from glass supports.

  • Made of 100% cotton fibre
  • Uniform and smooth
  • Produced using ultra-pure water
  • Contains no additives
Bestill nå

SPESIFIKASJONER

Nr. 703
Vekt 185 g/m²
Environmentally Preferable
Tykkelse 0,38 mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR