VWR®, Filter Tips for Rainin® LTS® Pipettes
Leverandør: VWR Collection
732-0799EA 1900 NOK
732-0799 732-0800 732-0801 732-1483
VWR®, Filter Tips for Rainin® LTS® Pipettes
Pipettespisser
These sterile aerosol filter tips are made of clear, pre-tested PP resin and fit Rainin® LTS® pipettes.
  • Fit and eject with significantly less force than most conventional pipette tips
  • Ultra-fine points and super-clear resin for excellent viewing of sample
  • Free of RNase, DNase, endotoxins, human DNA, protease and trace metals
  • Tested for ATP - Sustainable design thanks to use of virgin resins
Unlike pre-sterilised pipette tips from other brands, these tips use a validated sterile process (ISO) and can be used for medical research and diagnostics.