Manual compression sealer for AxyMat™ sealing mats, Axygen® EASY PRESS
Leverandør: Corning
IT-EP-R
732-0907EA 32575 NOK
732-0907
Manual compression sealer for AxyMat™ sealing mats, Axygen® EASY PRESS
Filmapplikatorer Sealing Film Applicators
Axygen® EASY PRESS is a manual device which provides simple and effective application of Axymat™ and/or Impermamat™ sealing mats to 24, 48, 96 or 384 well microplates, deep well plates, microtitre tube systems and PCR plates. The EASY PRESS can be quickly adjusted to handle any size plate of varying heights (384 PCR to 2 ml deep well blocks) using the large thumbwheel.