Sealing film roller, Axygen®
Leverandør: Corning
PCR-SP-ROLLER
732-1380EA 800 NOK
732-1380
Sealing film roller, Axygen®
Filmapplikatorer Sealing Film Applicators
Axygen® sealing film roller is a durable and efficient roller for applying sealing films to any plate. Sturdy plastic handle and semi-hard roller head are ideal for complete and easy film application.