Electrophoresis power supplies, EVT 300
Leverandør: EDVOTEK
509
732-1424EA 3650 NOK
732-1424
Electrophoresis power supplies, EVT 300
Strømkilder
EVT 300 power supplies can operate M6Plus units, two M12 units or two HexaGels (at 75 V). The EVT 300 runs gels as fast as 20 to30 minutes (at 150 V).