Horizontal electrophoresis apparatus, M36 HexaGel™
Leverandør: EDVOTEK
515
732-1425EA 5200 NOK
732-1425
Horizontal electrophoresis apparatus, M36 HexaGel™
Elektroforesesystemer Horisontal gelelektroforesesystemer
Six groups of students can load their own individual gels and the six gels can be run together in 30 to 40 minutes. No need for gel taping and gels can be poured quickly and easily using the gel tray sealing rubber end caps.
  • Seamless, injection-moulded casing
  • Safety interlock cover
  • Corrosion-resistant platinum electrodes
  • Safety insulated electrical leads
  • Adjustable leveling feet