VWR®, PCR Workstation
Leverandør: VWR Collection
732-2541EA 39575 NOK
732-2541
VWR®, PCR Workstation
Avtrekksskap og kabinetter PCR-arbeidsstasjoner og -skap
The VWR® PCR Workstation is a PCR enclosure for PCR sample preparation and other sensitive protocols. It offers dual decontamination action by UV inactivation of airborne and surface-bound contaminants and, therefore, represents an ideal environment for PCR set up. Stainless steel housing, front and side panels made of 8 mm Makrolon® protects from UV irradiation.
  • Active decontamination of work surface during non working time by UV irradiation
  • Additional inactivation of aerosol-bound contaminants by shielded UV Air Recirculator during operation
  • Contaminant