Forseglingstape, for MicroWell™-plate, Nunc™
Leverandør: Thermo Fisher Scientific
236366
732-2610EA 1700 NOK
732-2610
Forseglingstape, for MicroWell™-plate, Nunc™
Forseglinger Filmer og folier til mikroplateforsegling
Polyester film with acrylic adhesive. Provides an effective seal for all MicroWell™ plates used in PCR and cell culture.
  • Resistant to temperatures between −70 and +100 °C
  • Minimum evaporation and well-to-well cross-contamination
Dimensions: 134×80 mm
Dimensions: 134×80 mm  