Well caps for U96 DeepWell™ plates, Nunc™
Leverandør: Thermo Fisher Scientific
276000 276002 276003 276005 276011
732-2668EA 2850 NOK
732-2668 732-2669 732-2670 732-2672 732-2673
Well caps for U96 DeepWell™ plates, Nunc™
Forseglinger Filmer og folier til mikroplateforsegling
Made from thermoplastic elastomer (TPE), except pre-slit silicone well cap. For sealing 96-well plates, particularly 1,3 ml and 2,0 ml U96 DeepWell™ plates. TPE well caps are resistant to DMSO, ethanol and methanol. Silicone well cap is solvent resistant, but is not suitable for 100% DMSO.
  • Suitable for plates being used for chemical and compound storage
  • Produces an efficient seal with minimum protrusion into each well
  • Pierceable and self-closing
  • Can be used at temperatures down to −80 °C (except silicone cap, −20 °C)
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact
Miljøsertifikat ISO 14001 (Certificate Registration No.CERT-IT000330) NAR Certificate of Retirement