Lagringsplate, Deep well, 96-brønner
Leverandør: THERMO MATRIX TECHNOLOGIES
AB-0781 AB-1178 AB-1056
732-4879EA 4625 NOK
732-4879 732-3338 MATRAB-1056
Lagringsplate, Deep well, 96-brønner
Mikroplater Deepwell mikroplater
Thermo Scientific™ Abgene™ polypropylene deep well plates offer storage security for assays, compound libraries or storing samples for either intermediate or long term use.
  • Compatible with a variety of automated liquid handling for high throughput workflows
Sertifiseringer: Manufactured in Class 100,000 cleanroom under ISO 9001 conditions. ANSI compliant.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat IRECS; ISO 14001 (Certificate Registration No. EMS 752603)