VWR®, Filter Tips, Next Generation Refill, Low Retention
Leverandør: VWR Collection
732-3350EA 1925 NOK
732-3350
VWR®, Filter Tips, Next Generation Refill, Low Retention
Pipettespisser
These aerosol filter tips, with their patented filters made from high molecular weight polyethylene, offer complete protection from sample carry-over. Ideal for PCR and DNA manipulation.
  • Extremely low surface tension significantly reduces sample loss and provides substantially higher reproducibility when working with critical media
  • Easy to use refill system - new pipette tip racks, inserts and transfer cover preventing tip wobbling during refill and keeping tips clean
  • 95% renewable materials - compostable tip base from organic non petroleum resin, carton