PCR Workstation HEPA
Leverandør: VWR Collection
732-3409EA 63650 NOK
732-3409
PCR Workstation HEPA
Avtrekksskap og kabinetter PCR-arbeidsstasjoner og -skap
The VWR® PCR Workstation HEPA is a PCR enclosure for PCR sample preparation with an HEPA Filter. It offers dual-decontamination action by UV inactivation of airborne and surface-bound contaminants, therefore, representing an ideal environment for PCR set up and other sensitive protocols. In addition, the HEPA filter with UV provides a barrier against dust, bacteria and mould.
  • Active decontamination of work surface during non working time by UV irradiation
  • Additional inactivation of aerosol-bound contaminants by shielded UV air recirculator during