Deep Well-lagringsplater, 96-brønners, 2,2 ml, Mark II
Leverandør: THERMO MATRIX TECHNOLOGIES
AB-0932
732-4910EA 3125 NOK
732-4910
Deep Well-lagringsplater, 96-brønners, 2,2 ml, Mark II
Mikroplater Deepwell mikroplater
PP. 96-brønners plater med runde brønner for oppbevaring og screening.
  • 2,2 ml working volume when sealed with adhesive or heat seals or 1,8 ml when sealed with 96-cap sealing mat
  • Conical well bottom for ease of sample retrieval
  • Raised rim design with support and stacking ring
Colour: Natural.
Colour: Natural