Storage plates, 96-well, V-bottomed
Leverandør: THERMO MATRIX TECHNOLOGIES
AB-1058
732-4942EA 4075 NOK
732-4942
Storage plates, 96-well, V-bottomed
Mikroplater Deepwell mikroplater
PP.
  • V-bottomed well design for optimal sample recovery
  • Reinforced skirt for robotic manipulation
  • 0,2 ml maximum well volume