Protease inhibitor cocktail, FOCUS™ ProteaseArrest™
Leverandør: G Bioscienes
786-108F
733-1945EA 5900 NOK
733-1945
Protease inhibitor cocktail, FOCUS™ ProteaseArrest™
Cellekultursupplementer og tilsetninger
A ready to use, 100X concentrated, broad range protease inhibitor cocktail that is fully compatible with 2D electrophoresis and subsequent mass spectrometry.
  • Compatible to 2D gel electrophoresis
  • Broad spectrum protease inhibitor cocktail
  • 100X concentrated, ready to use solution
  • Inhibition of protease activity in protein preparations
  • Suitable for 2D gel sample preparation for protection of protein samples